Learn More
Sorbent Technologies Inc C18 Silica HPTLC Plates, w/UV254, 100% silanization, glass backed, 200um, 10x10cm, 25/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Sorbent Technologies Inc 2514156
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This high performance thin layer chromatography (HPTLC) plate comprises a C18 Silica layer. The Nano-Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 2-10um, and pH stability of 2-10. The TLC plates contain UV254 fluorescent indicator. The surface has been 100% treated with an octadecyl silane so that it is a reversed phase material. This analytical hard layer is bound to a glass support. It has a thickness of 200um and dimensions of 10x10cm. There are 25 plates per pack. Applications include: Alkaloids, amino acids, preservatives, optical brighteners, barbiturates, polycyclic aromatic hydrocarbons (PAH), drugs, peptides, flavonoids, phenols, indole derivatives, steroids
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.