Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1-C6 n-Paraffins, 15 ppm Each Component in Nitrogen, Analytical Standard, MilliporeSigma™ Supelco™

Recommended for qualitative analysis.

$229.41

Specifications

Concentration 15 ppm each component in nitrogen
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Butane; Ethane; Hexane; Methane; Pentane; Propane
Physical Form Gas
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-4358
View Documents
MilliporeSigma Supelco
501778
4 L
Each of 1 for $229.41
Add to Cart
 
Description

Description

Specifications

Specifications

15 ppm each component in nitrogen
Analytical Standard
Butane; Ethane; Hexane; Methane; Pentane; Propane
Gas
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit