Learn More
Research Products International Corp C Shaped Lead Ring Flask Weight, Vinyl Coated, Open, 2 3/4 Inch Inner Diameter for 1000-4000ml Flasks, 2 Pounds
Supplier: Research Products International Corp 248859
"C" Shaped Lead Rings help stabilize glass and plastic containers. Rings fit around neck of flasks and eliminate floating in water baths or tipping on benchtops. Open ring design allows adjustment to fit wide vessels. Vikem vinyl coating protects glassware and is resistant to most acids and solvents.