Learn More
C.L. Sturkey Select Disposable Microtome Blades
Ideal for all general paraffin applications and cryostat use
$143.90 - $146.00
Specifications
For Use With (Equipment) | Microtome Blade |
---|---|
Disposable | Yes |
Quantity | 50 Pk. |
Catalog Number | Mfr. No. | Profile | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Profile | Price | Quantity & Availability | |||||
22-210-052
|
C.L. Sturkey
DT554S50 |
High |
Pack of 50 for $146.00
Case of 10 PK for $1,309.00 |
|
22-210-051
|
C.L. Sturkey
DT315S50 |
Low |
Pack of 50 for $143.90
Case of 10 PK for $1,288.00 |
![]() |
Description
- State-of-the-art grinding and coating techniques are specially designed to create an edge that produces superior sections with every cut
- Standard Dispenser holds 50 blades per pack
Specifications
Microtome Blade | |
Yes | |
50 Pk. |