Learn More
PATTERSON DENTAL COMPANY C&B-Metabond Quick Base, Bottle (10 ml)
Supplier: PATTERSON DENTAL COMPANY S398
C&B-METABOND adheres to most surfaces you encounter in dentistry (cast metal, amalgam, dentin, enamel, cementum, acrylic or composite). Use the system to make fast intraoral ceramic repairs. C&B-METABOND chemically grafts to any composite.Strong, durable, reliable. Bonds several types of materials securely. Backed by a longer clinical history than any other adhesive cement in the world. Repair crowns that have loosened, short posts, broken teeth, porcelain pop-offs, etc.Extremely versatile – ideal for dental emergency procedures. Quick! formula features a longer working time and shorter set time, optimizing efficiency.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.