Learn More
Bachem H-Ala-Ala-Gln-OH
Supplier: Bachem 4026080.1000
H-Ala-Ala-Gln-OH, 290312-62-8, C11H20N4O5, Mr 288.30, 1g. By-product formed during the synthesis of the dipeptide Ala-Gln, glutamine source in parenteral nutrition.