Learn More
Chemglass Life Sciences BVC Control Aspirator System, 4L Polypro Bottle
SDP
Supplier:  Chemglass Life Sciences CLS900200
Encompass_Preferred
BVC Control Aspirator System, 4L Polypro Bottle. Chemglass Life Sciences is pleased to introduce the new line of BVC fluid aspirator systems for cell culture. They provide the flexibility to accommodate your decontamination protocol whether you autoclave or disinfect in the bottle. Control Model:The BVC control unit features a powerful, whisper-quiet Vacuubrand chemistry designed vacuum pump, thereby eliminating the need for central vacuum supply. The pump is turned on and off automatically on demand when using the hand controller. An easy to clean touch panel adjusts and indicated the vacuum level switch point. This provides the exact power you need, when you need it. Available with a 4 liter polypropylene bottle or 2 liter glass collection bottle.
Catalog No. 50-131-1384
Explore available promotions