Learn More
Phoenix S&T Inc Butterfly Portfolio Heater (20 cm x 6 cm)
SDP
Supplier:  Phoenix S&T Inc PSTBPH20W
Encompass
Even, stable(to within 0.2 ºC) temperature throughout the whole hot “pocket” formed by the closed Butterfly heater when controlled by Phoenix S&T’s column heater controller: PST-CHC. Easy placement of chromatographic fittings and columns, including vented columns, in the portfolio when it is “open”, and the closed heater can be placed adjacent to the MS source. Contents of the heater can be conveniently secured in place by heat-resistant tape. Maximum temperature achievable is 100 ºC. Accessories for insulating the union or Tee for high voltage liquid junction inside the heater are available.
Catalog No. NC1600026