Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butane-Powered Micro Burners
Provide safe, portable flame applications in a variety of settings.
Supplier: Wall Lenk Corporation 43
This item is not returnable.
View return policy
Description
Self-contained and require no wicks to replace or gas lines to install.
- Automatically ignite
- Tip-resistant wide base
- Refillable
- Reach temperature of up to 1315.6°C (2400°F)
- Work continuously for one hour on maximum setting
Specifications
Micro Burner | |
Angled Flame | |
1315.6°C |
Butane | |
Provide Safe, Portable Flame Applications in a Variety of Settings |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction