Learn More
Cole-Parmer Humboldt H-6225 Accuflame Straight Tube Gas TIRrill Bunsen Burner, Lp Gas
Supplier: Cole-Parmer 3613012
Straight Tube Gas Burners feature adjustable flames for very slow heating or quick boiling. Flame retainer prevents flame lifting or extinguishing. Tapered serrated gas inlet connections for easy tube mounting.