Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Burn-up Bin™ Biohazard Waste Box PROMO
Store flat, assemble in seconds.
Description
- Floor model box features integral handles for safety in use and transport
- Corrugated cardboard - single use
- 2 mil polyliner safely holds biohazard waste
- Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
- Graphics and biohazard symbols readily identify contents, encourage proper use
- Replacement bags are not available for the Burn-Up Bin as boxes are to be incinerated after use
Specifications
Floor model | |
5.25 in. | |
White | |
69 cm | |
Floor | |
Polyethylene | |
12 in. | |
2 mm | |
Cardboard |
13.3 cm | |
11 gal. | |
12 x 12 x 27 in. (30 x 30 x 69 cm) | |
30 cm | |
30 cm | |
Quick identification for non-hazardous waste paper products | |
12 in. | |
27 in. | |
Biohazard Waste Box |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction