Learn More
Hach Company Buret Kit, Automatic, Self-Zeroing
Supplier: Hach Company 2073900
Economical kit turns any 25 or 50 mL buret into an auto-filling, self-zeroing titration work station. Can be used with most aqueous titrants. Kit includes 1000 mL low-density polyethylene reservoir, 48” of tubing, connectors, reservoir tubing and extra closure without tubing hole for storing titrants or carrying to the field.