Learn More
Ace Glass, Inc. Buret, auto, 50mL, 29/42, 1:5 PTFE plug, complete with reservoir and pressure bulb/release tube, 1/10mL subdivisions
SDP
Supplier:  Ace Glass, Inc. 573546
Encompass_Preferred
Catalog No. 50-972-35