Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Bumper Guard For Volumetric Flask
SDPSCI_ED

These guards help prevent volumetric glassware from breaking when tipping over.

$12.04 - $43.66

Specifications

For Use With (Equipment) Volumetric Flask
Product Type Bumper Guard
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Media Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Media Price Quantity & Availability  
S16070
SDPSCI_ED
View Documents
United Scientific™
BMP250PK5
250 mL
Pack of 5 for $43.66
Add to Cart
 
S15908
SDPSCI_ED
View Documents
United Scientific™
BMP100PK5
100 mL
Pack of 5 for $23.86
Add to Cart
 
S15965
SDPSCI_ED
View Documents
United Scientific™
UNBMP500
500 mL
Each for $12.04
Add to Cart
 
S15741
SDPSCI_ED
View Documents
United Scientific™
UNBMP1000
1000 mL
Each for $16.20
Add to Cart
 
S15886
SDPSCI_ED
View Documents
United Scientific™
BMP25PK5
25 mL
Pack of 5 for $17.40
Add to Cart
 
Description

Description

  • Silicone bumpers slip easily onto any volumetric flask. In the event of a flask being accidentally tipped over, the wide bumpers cushion the neck from hitting the table surface.
  • Color coded and available in five different sizes.
  • Wide design to cushion flasks if tipped over
  • Made of high-quality silicone
Specifications

Specifications

Volumetric Flask
Bumper Guard
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit