Learn More
Fine Science Tools Bulldog Serrefines, Curved, Stainless Steel, 28 mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1805128
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Product: Bulldog Serrefine - Curved/28mm/ Shape: Curved/ Jaw Dimensions: 9 x 1.6mm/ Clamping Pressure: Medium/ Alloy: Stainless Steel/ Material: / Length: 28mm/ Serrated: Yes/ Unit: Each