Learn More
Bullard™ EDP10 Electric Driven Free-Air™ Pump
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Provides continuous airflow for up to two people.
Supplier: Bullard™ EDP10
Description
- Supplies air for one hood or two full facemask airline respirators
- Weighing only 49 lbs., this pump can be easily transported from one job site to another
- Meets UL requirements and CSA approved
Specifications
EDP10 | |
60 lb. | |
Electric Driven Air Pump |