Learn More
Brew Plate™ Bulk Sleeve - Funkadelic Quad Plate
Brew Plate™ Bulk Sleeve - Funkadelic Quad Plate is for the detection of all yeasts and bacteria found in your brew
Supplier: Brew Plate™ BREWPLTBULKFNQP
Description
Each quadrant is made up of different media that detects various species:
- Quadrant 1: Wallerstein Nutrient Media (WLN) is non-selective and detects all wild yeast, pitching yeast and some bacterial strains that are common in brew
- Quadrant 2: Lee's Multi-Differential Agar (LMDA) detects lactic acid producting bacteria including Lactobacillus and Acetobacter species. Acid producers are easily detected by a change in the media color from green to yellow
- Quadrant 3: Hsu's Lactobacillus/Pedioccocus Agar (HLP) detects Lactobacillus and Pediococcus bacteria. For optimal growth, an anaerobic gas generator sachet should be used during incubation
- Quadrant 4: Lin's Cupric Sulfate Media (LCSM) detects non-Saccharomyces wild yeast and inhibits the growth of brewer's yeast.
1 Free Sequence ID per sleeve
Specifications
Refrigeration | |
Agar | |
Sleeve |
United States | |
Bacteria, Yeast | |
25 Plates |
Safety and Handling
ShelfLife : 90 Days