Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl Bulk Shelving Shelf Divider

Shelf divider for use with bulk shelving systems.

$45.90 - $52.30

Specifications

For Use With (Application) Shelving
Includes Divider
Color Chrome
Material Steel
View More Specs

Products 2
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
11-222-142
View Documents
Wanzl
T.BCR24DV
24 in.
Each for $45.90
Add to Cart
 
11-222-143
View Documents
Wanzl
T.BCR36DV
36 in.
Each for $52.30
Add to Cart
 
Description

Description

  • Shelf divider accessory
  • For use with bulk shelving system
Specifications

Specifications

Shelving
Divider
Chrome
Steel
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit