Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Bulk Filtered Micropipette Tips
Designed to be used with variable or fixed single and multi channel micropipettes. Eisco™ Bulk Filtered Micropipette Tips include a hydrophobic polyethylene filter to protect samples and the micropipette from contamination.
Supplier: Eisco™ CH0591J
Description
- Autoclavable
- Contain hydrophobic polyethylene filter
- Free of lubricants, dyes and heavy metals
- Non-sterile
- Supplied in resealable plastic bag
Specifications
2 to 200 μL | |
Non-sterile | |
Autoclavable | |
Bulk | |
1000 | |
Universal Fit Micro | |
Polypropylene |
Filtered | |
Clear | |
Electrophoresis, PCR | |
Bag | |
Single-use | |
No |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction