Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Bulk Filtered Micropipette Tips
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Designed to be used with variable or fixed single and multi channel micropipettes. Eisco™ Bulk Filtered Micropipette Tips include a hydrophobic polyethylene filter to protect samples and the micropipette from contamination.
Supplier: Eisco™ CH0591I
Description
- Autoclavable
- Contain hydrophobic polyethylene filter
- Free of lubricants, dyes and heavy metals
- Non-sterile
- Supplied in resealable plastic bag
Specifications
2 to 100 μL | |
Non-sterile | |
Autoclavable | |
Bulk | |
1000 | |
Universal Fit Micro | |
Polypropylene |
Filtered | |
Clear | |
Electrophoresis, PCR | |
Bag | |
Single-use | |
No |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction