Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer Solution pH 7.0 at 25°C, Color Coded Yellow, Certified, LabChem™
Supplier: LabChem, Inc. LC1238020
Specifications
Buffer Solution pH 7.0 at 25°C | |
Yellow | |
7778-77-0, 1310-73-2 | |
7.00 ±0.01 | |
20 x 20 mL | |
KH2PO4 | |
Traceable to NIST | |
136.09 |
Color Coded Yellow | |
99.24, 0.68, 0.08 | |
Phosphorus oxides; Sodium oxide | |
Liquid | |
Passes Test | |
Pack | |
Soluble in water | |
Certified |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction