Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 4.0 at 25°C, Color Coded Red, Certified, LabChem™

Supplier:  LabChem, Inc. LC122806

 View more versions of this product

Catalog No. LC122806


Add to Cart

Description

Description

Specifications

Specifications

Buffer Solution pH 4.0 at 25°C
Red
877-24-7, 50-00-0, 67-56-1
1g/mL
Liquid
Passes Test
Cubitainer™
Traceable to NIST
204.23
Color Coded Red
98.935, 1.02, 0.04, 0.005
Formaldehyde; Carbon monoxide; Carbon dioxide
4.00 to 4.01
10 L
KHC8H4O4
1g/mL
Soluble in water
Certified
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit