Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pH Buffer Salt, pH 9.18, in ready to mix packets, Certified, Fisher Chemical
Click to view available options
Quantity:
12 Ea.
Packaging:
Envelopes
Description
- Each packet contains sufficient material to prepare 1 liter of solution with a pH of 9.18 ± 0.02 at 25°C
- Convenient and ready-to-mix
- Mix with deionized water to make 1L of pH buffer
- Eliminates the need to store or carry bottles of buffer solution - simply transport the ready-to-mix packets
- Intended for research use only; not for human or animal diagnostic purposes
Specifications
Melting Point | 75°C |
Boiling Point | 1575°C |
Color | White |
Density | 1.7300g/mL |
pH | 9.18 ±0.02 |
Physical Form | Powder or Solid |
Quantity | 12 Ea. |
Identification | Pass Test |
Packaging | Envelopes |
Grade | Certified |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction