Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C (Color Coded Green), Ricca Chemical
$56.60 - $1082.00
Specifications
CAS | 7558-79-4, 7778-77-0, 7732-18-5 |
---|---|
Color | Green |
CAS Max % | 100.0, 0.9, 0.5 |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
15514-16
|
Ricca Chemical Company
1551.416 |
500 mL | Natural Poly Bottle |
Each for $56.60
|
|
||||
15514-32
|
Ricca Chemical Company
1551.432 |
1 L | Natural Poly Bottle |
Each for $94.40
|
|
||||
15514-1CT
|
Ricca Chemical Company
1551.41CT |
4 L | Cubitainer |
Each for $199.20
|
|
||||
15514-1
|
Ricca Chemical Company
1551.41 |
4 L | Natural Poly Bottle |
Each for $190.90
|
|
||||
15514-25
|
Ricca Chemical Company
1551.42.5 |
10 L | Cubitainer |
Each for $289.50
|
|
||||
15514-5
|
Ricca Chemical Company
1551.45 |
20 L | Cubitainer |
Each for $398.50
|
|
||||
1551455
|
Ricca Chemical Company
1551.455 |
55 gal. | Poly Drum |
Each for $1,082.00
|
|
||||
Specifications
7558-79-4, 7778-77-0, 7732-18-5 | |
Green | |
100.0, 0.9, 0.5 | |
Liquid |
Safety and Handling
ShelfLife : 24 months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title