Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 4.000.01 (25°C), MilliporeSigma™ Supelco™

$59.00 - $295.00

Specifications

Color Red
MDL Number MFCD00130682
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-5468
View Documents
MilliporeSigma Supelco
B5020500ML
500 mL
Each for $59.00
Add to Cart
 
11-100-5430
View Documents
MilliporeSigma Supelco
B50206X500ML
6 x 500 mL
Each for $267.00
Add to Cart
 
11-100-5427
View Documents
MilliporeSigma Supelco
B50201GA
1 gal.
Each for $295.00
Add to Cart
 
Description

Description

Specifications

Specifications

Red
MFCD00130682
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit