Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Buffer Pad
3M™ Red Buffer Pad quickly cleans and removes scuff marks and enhances floor appearance.
$30.50 - $58.25
Specifications
Product Type | Buffer Pad |
---|---|
Color | Red |
For Use With (Application) | Spray Cleaning Protected Floors |
Material | Non-Woven Polyester Fiber |
Quantity | 5/Cs. |
Catalog Number | Mfr. No. | Diameter (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Price | Quantity & Availability | |||||
19-072-011
|
3M™
7000000677 |
13 in. |
Case of 5 for $30.50
|
|
|||||
19-072-004
|
3M™
7000000663 |
20 in. |
Case of 5 for $58.25
|
|
|||||
Description
- Use to light scrub without dulling the finish
- Can be used for everyday cleaning under and automatic scrubber
- Specifically designed for spray buffing
- Cleans when damp; Buffs when dry
- Use on rotary or automatic floor machines
- Works well on automatic floor scrubbers for light duty cleaning
Specifications
Buffer Pad | |
Spray Cleaning Protected Floors | |
5/Cs. |
Red | |
Non-Woven Polyester Fiber |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title