Learn More
Chemglass Life Sciences Bubbler, In-Line, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF051020
Bubbler, In-Line, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Bubbler has a "T" connection at the top to allow it to be mounted in-line while the reservoir head prevents oil from being sucked back into the system. Volume below the sidearm is approx 40mL. Bubbler has 8mm OD top and side tubulations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.