Learn More
UNITED CHEMICAL TECHNOLOGIES BSTFA W/ 1% TMCS 100GRBSTFA W
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: UNITED CHEMICAL TECHNOLOGIES SBSTFA1100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Selectra-sil BSTFA w/1% TMCS. O-bis(trimethylsilytrifluoroacetamide) w/ 1% Trimethylchlorosilane. 100 g vial.
This item is not returnable.
View return policy