Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Standard Grade, 1 kg
Supplier: Bioworld 220700086
BSA (Bovine Serum Albumin), Fraction VProtein with numerous biochemical applications including ELISAs, immunoblots, and immunohistochemistryHeat shock isolationSpecifications:pH (5% water): 6.5-7.5 Heavy Metals (Pb): <0.001% DNase: None detected RNase: None detected Protease: None detectedUsually ships within 24 hours. This product is sold for laboratory use only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.