Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromotris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Quantity | 5 g |
UN Number | UN1759 |
Sensitivity | Light sensitive |
Formula Weight | 388.07 |
Percent Purity | ≥98% |
Chemical Name or Material | Bromotris(dimethylamino)phosphonium hexafluorophosphate |
Safety and Handling
Hazard Category | H314-H335-H350 |
Hazard Statement | GHS H Statement H350-H314-H318 May cause cancer. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S. |
TSCA | No |
Recommended Storage | Keep cold |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction