Learn More
Biolegend Brilliant Violet 605TM anti-mo
Supplier:  Biolegend 127025
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD317 (BST2, PDCA-1) [927]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-272-2706
Explore available promotions