Learn More
Research Products International Corp Bremshield, Vertical Bench-Top Beta Shield, 3/8 Inch Thick, 15 x 21 3/4
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp BR201
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Vertical shields feature a front mounted anti-tip stop to provide maximum stability and a dual angled window for distortion free viewing of work area.Can be used in combination with horizontal beta shields, sold separately, to configure a personal and enclosed work station.