Learn More
Electron Microscopy Sciences Breathe-Easy for Petri Dishes
Supplier: Electron Microscopy Sciences 7053619
Gas-permeable polyurethane membrane for petri dishes are in pre-cut strips. Cell growth is unimpeded since the strips allow for beneficial exchange of oxygen and carbon dioxide for cellular growth drying. The strips acts as a microbial barrier that prevents microbial and vial contamination during growth and storage of cells.
Strip design allows film to be protected during application; a tab remains attached for easy removal.
Dishes sealed with Breathe-Easy strips may be incubated and/or stored and ambient temperatures, refrigerated or frozen (-80 degree C to 100 degree C).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.