Learn More
Branson Ultrasonics™ Liquid Rust Stripper
For derusting and descaling.
Supplier: Branson Ultrasonics™ CPN955003
Description
Branson™ Liquid Rust Stripper is used for derusting and descaling of ferrous metals. Excellent for use with ultrasonics for cleaning of automotive, aero-space, components, and removal of carbonized and greasy soils in CIP applications, cleaning of food processing equipment and bottle washing.Specifications
For derusting and descaling of Ferrous Metals | |
1 gal. |