Learn More
Branson Ultrasonics™ Bransonic™ Metal Cleaners
For ultrasonic cleaners
Supplier: Branson Ultrasonics™ 100955826
Specifications
MC1 Metal Cleaner | |
Used to remove Oils, Greases and soils from Aluminum and Aluminum alloys, Copper, Brass and Steel substrates | |
19 L |
Liquid | |
5 gal. |