Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Branson Ultrasonics™ Bransonic™ Metal Cleaners
For ultrasonic cleaners
Supplier: Branson Ultrasonics™ 100955834
Specifications
MC2 Metal Cleaner | |
12.5 | |
100°C | |
Used to remove Oils, Greases and soils from Ferrous Metals, Steel, Copper, Titanium Alloys, Stainless Steel, for cutting lubricants | |
1 gal. |
Liquid | |
Clear Yellow | |
7 to 10% | |
MC-2 Metal Cleaner | |
3.78 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction