Learn More
Branson Ultrasonics™ Bransonic™ Metal Cleaners
For ultrasonic cleaners
Supplier: Branson Ultrasonics™ 100955824
Specifications
MC1 Metal Cleaner | |
9.4 | |
100°C | |
Used to remove Oils, Greases and soils from Aluminum and Aluminum alloys, Copper, Brass and Steel substrates |
Liquid | |
Clear Amber | |
7 to 10% | |
1 gal. |