Learn More
BrandTech™ BRAND™ Dispensette™ III Digital and Analog Bottle-Top Dispensers
For safe and accurate dispensing of many laboratory liquids, including most acids, bases, salt solutions, scintillation fluids and many organic solvents
Supplier: BrandTech™ 4701141
Includes: 45mm thread, discharge tube, telescoping filling tube, valve mounting tool, polypropylene thread adapters (10mL or smaller units supplied with 24, 28, 33 and 38mm adapters; 25 and 50mL units come with 28, 33 and 38mm adapters; 100mL units come with 33 and 38mL adapters), certificate of performance, and operating manual
Description
Low-friction floating piston design provides reliable, smooth, one-handed operation without wiping seals to wear out. Suitable for most laboratory applications and fit most bottles with included adapters. Autoclavable for use with sterile reagents.
- Digital Easy Calibration™ and analog models from 0.05 to 100mL
- Rugged floating piston design adapts to nearly any unpressurized container
- All models can be calibrated easily
- Digital models have mechanical digital displays and include Easy Calibration feature
- Integral SafetyPrime™ valve permits recirculation of dispensed liquid into reagent bottle during priming to reduce sputtering and conserve reagents
- Safety discharge system protects against inadvertent dispensing
- Polypropylene cylinder sleeve helps contain dispenser contents in event of breakage
- Flowpath consists of: borosilicate glass, Al2O3 ceramic, platinum-iridium, PFTE, ETFE, FEP and PFA
- Polypropylene safety screw cap on discharge tube
- Accuracy for all models is within ±≤0.5% (±≤1.0% for 0.05-0.5mL size), with reproducibility of ≤0.1% (≤0.2% for 0.05-0.5mL size)
- Autoclavable at 250°F (121°C) for sterile dispensing or decontamination
- Supplied with thread adapters and telescoping filling tube that adjusts to length without cutting
- Valve block rotates 360°
For dispensing concentrated fuming acids, peroxides, or difficult organic solvents, select the Dispensette Organic. Not for use with hydrofluoric acid (HF) or carbon disulfide (CS2).
Order Info
Accessories available that allow dispenser to adapt to nearly any unpressurized reagent container; tube-to-tube serial dispensing with flexible discharge tubing, and dispense sterile or moisture/CO2 sensitive reagents. For a complete listing, visit www.fishersci.com.
Compliance
Can be calibrated by users for GLP and ISO 9000 compliance.
Specifications
≤0.5% ±50μL | |
Autoclavable | |
Up to 2.2 g/cm3 | |
1 to 10 mL | |
SafetyPrime™ valve, 45mm thread, discharge tube, 125 to 240mm L telescoping filling tube, valve mounting/calibration tool, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual | |
600 mbar | |
59°F to 104°F | |
500mm2/sec. |
Dispensette III Bottletop Dispenser | |
GLP, ISO 9000 | |
Borosilicate Glass | |
Analog | |
0.2 mL | |
≤0.1% 10μL | |
15°C to 40°C | |
1 Year |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.