Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ BradyPrinter M611 Mobile Label Printer and Brady Workstation PWID Software Suite
Get the complete print-from-anywhere solution with this kit that features the BradyPrinter M611 Mobile Label Printer and Brady Workstation Safety & Facility Identification software.
Supplier: Brady™ M611AMBWSPWID
Description
- Easily create and edit labels in the field and print directly from your mobile device via Bluetooth or Wi-Fi
- Work all day with the rugged design, long-lasting battery and built-in auto cutter
- Winner of EC&M's Product of the Year 2019 in the Labeler or Printer category and CI&M's silver innovator award in the cabling media category
- Brady Workstation Product and Wire ID Software Suite makes creating product identification and wire marking labels fast and efficient
Specifications
Thermal Transfer | |
Monochrome | |
(1) AC Adapter/Battery Charger (M-AC-143110); (1) BradyPrinter M611 Mobile Printer (M611-AM-BT-W), (1) Brady WorkStation Product and Wire Identification Software Suite, (1) Hard Case; (1) Li-Ion Internal Rechargeable Battery; (1) Matte White Polyester General Identification Labels (PTL-97-488); (1) R4300 Series Printer Ribbon (M61-R4310); (1) USB Cable (BMP410USB); (1) USB Flash Drive with Product Info and Windows Driver | |
1.33 in./sec. | |
Brady™ Workstation; Brady™ Workstation Mobile Apps | |
PC Connect | |
Data and Telecommunications labeling, Facility identification, Laboratory labeling, Safety identification, Wire and Cable labeling | |
110 to 240 V |
10.97 x 4.46 x 4.55 in. | |
320 x 240 pixels | |
256 MB Flash, 512 MB RAM | |
320 x 240 | |
Bluetooth; USB; USB 2.0; Wi-Fi | |
FCC Part 15 Class A | |
2 Years |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction