Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Key Retaining Keyed Different Steel Padlocks with 2 inch Shackle Locks

Key retaining safety feature prevents key from being removed until closed

Supplier:  Brady™ 123282

 View more versions of this product

Catalog No. 19-103-425

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Explore available promotions

Includes: 1 key

Description

Description

  • Key-retaining steel padlock has a 2 in. reinforced laminated steel shackle
  • Hardened steel body withstands severe physical abuse and tampering
  • Offers individually coated body plates that provide superior rust protection and corrosion resistance
  • Available in a variety of different colors
Specifications

Specifications

Brown
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.