Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Electrical Hazard Signs: DANGER - HIGH VOLTAGE OVERHEAD
Identify electrical hazard and high voltage areas
$30.75 - $145.00
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145, PSTC-33, |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | DANGER High Voltage Overhead |
Catalog Number | Mfr. No. | Height (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Price | Quantity & Availability | |||||
19-035-897
|
Brady™
69024 |
14 in. |
Each for $145.00
|
|
|||||
19-035-900
|
Brady™
84895 |
7 in. |
Each for $30.75
|
|
|||||
Description
- Available in two materials - fiberglass and polyester
- Available in combination of two different widths and heights
- Black/red on white
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33, | |
Surface Mount | |
DANGER High Voltage Overhead | |
Red on White |
1 Ea. | |
-40°F | |
Electrical Hazard Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title