Learn More
Brady™ Cleat for Clamp-on Breaker Lockouts
Cleat for Clamp-on Breaker Lockouts are 480/600V lockout cleats to safely lock out a wide range of applicable breakers.
Supplier: Brady™ 65406
Safety Services Available
Overview
LOTO Assessments
Description
- Cleats easily clip onto the bottom of clamp-on breaker lockout devices.
- Assures positive restraint for breakers with long, sliding switch throws
- Package includes cleat accessory only (lockout device sold separately)
- Extremely versatile - works on a wide range of single-pole and internal-trip multi-pole breakers
Specifications
Lockout Cleat | |
Red | |
480/600V lockout, clamp-on breaker lockout |