Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Braco Manufacturing Lens Cleaning Fluids

Click to view available options
Description:
Fog-Be-Gone
Pure Sight
Quantity:
2 oz.
16 oz.
Includes:
Flip Top
Spray pump
Specifications
Description | Fog-Be-Gone |
Product Type | Cleaning solution |
Includes | Flip Top |
Quantity | 2 oz. |