Learn More
Ampco Safety Tools Box End Wrench (Imperial)
AMPCO™ Box End Wrench performs in high-risk and demanding environments.
$208.50 - $2880.00
Specifications
Product Type | Box End |
---|---|
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
19-173-447
|
Ampco Safety Tools
4600 |
1/2 in. |
Each for $208.50
|
|
|||||
19-173-448
|
Ampco Safety Tools
4602 |
9/16 in. |
Each for $208.50
|
|
|||||
19-173-449
|
Ampco Safety Tools
4604 |
19/32 in. |
Each for $208.50
|
|
|||||
19-173-450
|
Ampco Safety Tools
4606 |
5/8 in. |
Each for $228.50
|
|
|||||
19-173-451
|
Ampco Safety Tools
4608 |
11/16 in. |
Each for $228.50
|
|
|||||
19-173-452
|
Ampco Safety Tools
4610 |
3/4 in. |
Each for $286.00
|
|
|||||
19-173-453
|
Ampco Safety Tools
4612 |
25/32 in. |
Each for $286.00
|
|
|||||
19-173-454
|
Ampco Safety Tools
4614 |
13/16 in. |
Each for $324.50
|
|
|||||
19-173-455
|
Ampco Safety Tools
4616 |
7/8 in. |
Each for $324.50
|
|
|||||
19-173-456
|
Ampco Safety Tools
4618 |
15/16 in. |
Each for $382.50
|
|
|||||
19-173-457
|
Ampco Safety Tools
4620 |
31/32 in. |
Each for $382.50
|
|
|||||
19-173-458
|
Ampco Safety Tools
4622 |
1 in. |
Each for $382.50
|
|
|||||
19-173-459
|
Ampco Safety Tools
4624 |
1-1/16 in. |
Each for $419.50
|
|
|||||
19-173-460
|
Ampco Safety Tools
4626 |
1-1/8 in. |
Each for $419.50
|
|
|||||
19-173-461
|
Ampco Safety Tools
4628 |
1-3/16 in. |
Each for $426.50
|
|
|||||
19-173-462
|
Ampco Safety Tools
4630 |
1-1/4 in. |
Each for $470.50
|
|
|||||
19-173-463
|
Ampco Safety Tools
4632 |
1-5/16 in. |
Each for $528.50
|
|
|||||
19-173-464
|
Ampco Safety Tools
4634 |
1-3/8 in. |
Each for $528.50
|
|
|||||
19-173-465
|
Ampco Safety Tools
4636 |
1-7/16 in. |
Each for $528.50
|
|
|||||
19-173-466
|
Ampco Safety Tools
4638 |
1-1/2 in. |
Each for $528.50
|
|
|||||
19-173-467
|
Ampco Safety Tools
4640 |
1-9/16 in. |
Each for $653.00
|
|
|||||
19-173-468
|
Ampco Safety Tools
4642 |
1-5/8 in. |
Each for $653.00
|
|
|||||
19-173-469
|
Ampco Safety Tools
4644 |
1-11/16 in. |
Each for $653.00
|
|
|||||
19-173-470
|
Ampco Safety Tools
4646 |
1-3/4 in. |
Each for $766.50
|
|
|||||
19-173-471
|
Ampco Safety Tools
4648 |
1-13/16 in. |
Each for $766.50
|
|
|||||
19-173-472
|
Ampco Safety Tools
4650 |
1-7/8 in. |
Each for $766.50
|
|
|||||
19-173-473
|
Ampco Safety Tools
4652 |
2 in. |
Each for $972.50
|
|
|||||
19-173-474
|
Ampco Safety Tools
4654 |
2-1/16 in. |
Each for $972.50
|
|
|||||
19-173-475
|
Ampco Safety Tools
4656 |
2-1/8 in. |
Each for $1,296.00
|
|
|||||
19-173-476
|
Ampco Safety Tools
4658 |
2-3/16 in. |
Each for $1,296.00
|
|
|||||
19-173-477
|
Ampco Safety Tools
4660 |
2-1/4 in. |
Each for $1,296.00
|
|
|||||
19-173-478
|
Ampco Safety Tools
4662 |
2-5/16 in. |
Each for $1,625.00
|
|
|||||
19-173-479
|
Ampco Safety Tools
4664 |
2-3/8 in. |
Each for $1,625.00
|
|
|||||
19-173-480
|
Ampco Safety Tools
4666 |
2-7/16 in. |
Each for $1,625.00
|
|
|||||
19-173-481
|
Ampco Safety Tools
4668 |
2-1/2 in. |
Each for $1,819.00
|
|
|||||
19-173-482
|
Ampco Safety Tools
4670 |
2-9/16 in. |
Each for $1,819.00
|
|
|||||
19-173-483
|
Ampco Safety Tools
4672 |
2-3/4 in. |
Each for $2,060.00
|
|
|||||
19-173-484
|
Ampco Safety Tools
4674 |
2-15/16 in. |
Each for $2,320.00
|
|
|||||
19-173-485
|
Ampco Safety Tools
4676 |
3 in. |
Each for $2,320.00
|
|
|||||
19-173-486
|
Ampco Safety Tools
4678 |
3-1/8 in. |
Each for $2,880.00
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Box End | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.