Learn More
BIOSEARCH TECHNOLOGIES INC NP-BSA (Bovine Serum Albumin), Ratio 1-4, 10mg
Supplier: BIOSEARCH TECHNOLOGIES INC N5050XL10
Appearance: yellowish powder, Absorption Maximum (Lambda Max): 430, Extinction Coefficient at Lambda max: 4230. 4-Hydroxy-3-nitrophenylacetyl hapten is conjugated to BSA (Bovine Serum Albumin) protein through lysine by amide bonds.