Learn More
Vector Laboratories Bovine Serum Albumin (BSA), Immunohistochemical Grade 500 mg
SDP
Supplier:  Vector Laboratories SP5050500
Encompass_Preferred
This ultrapure grade of bovine serum albumin (BSA) can be used as a diluent or a blocking agent. It is free of the impurities often present in other grades of BSA that can introduce artifacts or increase background staining in immunohistochemical assays, ELISAs, or blot-based protein detection.
Catalog No. NC9639624