Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 10 g
Supplier: Sigma Aldrich Fine Chemicals Biosciences A703010G
Heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%, Synonym: Albumin bovine serum, BSA, Bovine albumin. Bovine Serum Albumin has been used: as a constituent of the medium used for the differentiation of human pluripotent stem cells, in enzyme-linked immunosorbent assay (ELISA) and sandwich ELISA, and to permeabilize fetal cortical cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.