Learn More
MP Biomedicals™ Casein Hydrolysate Enzymatic, Powder
Casein hydrolysate enzymatic is recommended for use as Animal diet
$177.40 - $602.50
Specifications
Product Type | Animal Diet |
---|---|
Format | Powder |
For Use With (Application) | Animal diet |
Storage Requirements | Room Temperature |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP210129001
|
MP Biomedicals™
0210129001 |
1 lb. |
Each for $177.40
|
|
|||||
MP210129005
|
MP Biomedicals™
0210129005 |
5 lb. |
Each for $602.50
|
|
|||||
Description
Applications:
Casein hydrolysate enzymatic is recommended for use as Animal diet.
Product Description:
Casein hydrolysate enzymatic is a pancreatic hydrolysate of casein. This hydrolysate contains, in the form of mixed amino acids and peptides, all amino acids originally present in casein. It is a refined product providing high quality soluble source of amino acids, polypeptides and peptides.
Specifications
Animal Diet | |
Powder | |
Animal diet | |
Room Temperature |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.