Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radians Bouton Lens Cleaners

missing translation for 'orderingAttributeHoverText'
Product Type:
Cleaning solution
Includes:
Flip-top container
Twist cap
Quantity:
2 oz.
16 oz.
Specifications
Certifications/Compliance | OSHA |
Description | Lens cleaner |
Product Type | Cleaning solution |
For Use With (Application) | Eliminate static with an anti-fog solution |
Includes | Flip-top container |
Quantity | 2 oz. |
Silicone Free | Yes |