Learn More
PIP™ Bouton Contempo Goggles
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Includes:
Wide elastic headband with adjustable buckle
Wide neoprene headband with adjustable buckle
Description
- Wraparound lens design with distortion-free panoramic viewing
- Ideal for general manufacturing and anywhere eye protection is needed.
- Offers indirect ventilation for liquid splash protection
- Wide facial flange for a secure seal
- Nose area split to fit the vast majority of users
- Clear blue transparent frame to prevent yellowing
- Fits comfortably over most prescription glasses
- Headband strap attaches to frame with a swivel clip to ensure a comfortable fit
- 6 Base curve lens
- Meets ANSI Z87.1
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Frame Color | Light Blue |
Includes | Wide neoprene headband with adjustable buckle |
Warning - Prop 65 - Reproductive Harm